Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLK-1 Rat anti-Mouse, Unlabeled, Clone: AVAS 12A1, BD

Catalog No. BDB550549 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550549 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550549 Supplier BD Biosciences Supplier No. 550549
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The Avas 12α1 antibody reacts with fetal liver kinase 1 (Flk-1), a receptor protein tyrosine kinase closely related to CD117 (c-kit) and CD140a (PDGF Receptor a chain) of the immunoglobulin superfamily. Flk-1, also known as VEGF Receptor-2 (VEGF-R2)

Host Species: Rat
Clone: AVAS 12α1
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse Flk1 and human IgG1 fusion protein

Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen FLK-1
Applications Flow Cytometry
Classification Monoclonal
Clone AVAS 12A1
Concentration 31.25μg/mL
Conjugate Unconjugated
Description Fetal liver kinase 1; CD309; Kdr; VEGF receptor-2; VEGFR-2
Formulation Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Flk1 and human IgG1 fusion protein
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.