Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flotillin-1 Mouse, Unlabeled, Clone: 18, BD

Catalog No. BDB610820 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610820 50 μg
BDB610821 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610820 Supplier BD Biosciences Supplier No. 610820
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Caveolae are specialized membrane invaginations of 50-100 nm present in all cells, but abundant in endothelium, muscle cells, and adipocytes. These plasma membrane microdomains function in transcytosis of macromolecules, and are the sites of potocytosis, where small molecules are concentrated and transferred inside the cells by glycosylphosphatidylinositol (GPI)-linked receptors. Caveolin, a 22kDa protein and a well known marker for these plasma membrane microdomains, plays a structural role in these specializations. Flotillin-1 was isolated from the Triton X-100 insoluble buoyant fraction, characteristic of caveolae. Although the mRNA expression of both Flotillin-1 and Caveolin
is very similar, Caveolin is undetectable in brain, while Flotillin-1 is very abundant. Flotillin-1 is a close homolog of the Epidermal Surface Antigen (ESA/Flotillin-2), which also colocalizes in the caveolae. Thus, Flotillin-1 and its relative ESA/Flotillin-2 are now incorporated into the expanding list of proteins co-localized at the caveolae which includes PKCα, Ras, Rap Src-like kinases, Gαβγ,and GPI-linked receptors.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Flotillin-1
Applications Western Blot
Classification Monoclonal
Clone 18
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse Flotillin aa. 312-428
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.