Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flotillin-2 Mouse, Unlabeled, Clone: 29, BD

Catalog No. p-3855936 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610383 50 μg
BDB610384 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610383 Supplier BD Biosciences Supplier No. 610383
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Flotillin-2/ESA (epidermal surface antigen) plays a role in intercellular epidermal adhesion. Epidermal surface proteins have an important role in normal epidermal maintenance and function. In addition, they are involved in various autoimmune diseases in which auto-antibodies bind to normal epidermal cell surface proteins. This binding causes cell disadherence that leads to blister formation. Such antibodies have been utilized to identify new proteins involved in epidermal cell adhesion. One of these antibodies, ECS-1, was used to isolate Flotillin-2/ESA. The single copy ESA gene is located on the long arm of human chromosome 17, in the region containing the gene for neurofibromatosis 1 (NF1). ESA mRNA is expressed in cultured keratinocytes, melanocytes, fibroblasts, and carcinoma and melanoma cell lines. Flotillin-2/ESA may play a critical role in epidermal structure and maintenance.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Flotillin-2
Applications Western Blot
Classification Monoclonal
Clone 29
Concentration 250μg/mL
Conjugate Unconjugated
Description ESA
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.