Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescent Gold Rabbit anti-All, Polyclonal, MilliporeSigma™

Catalog No. ab153imi Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB153IMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB153IMI Supplier MilliporeSigma Supplier No. AB153I
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Fluorescent Gold Antibody is a highly specific rabbit polyclonal antibody, that targets Fluoro-Gold and has been tested in immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fluorescent Gold
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Host Species Rabbit
Immunogen KLH-conjugated Fluorescent Gold.
Purification Method Protein A purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) N/A
Target Species All species
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.