Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FoxO1/FKHR Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5939
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5939 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5939 Supplier R&D Systems Supplier No. MAB5939
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FoxO1/FKHR Monoclonal specifically detects FoxO1/FKHR in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen FoxO1/FKHR
Applications Immunocytochemistry
Classification Monoclonal
Clone 597554
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q12778
Gene Alias FKH1, FKHR, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma
Gene Symbols FOXO1
Host Species Mouse
Immunogen E. coli- derived recombinant human FoxO1/FKHR Ala353-Gly655 Accession # Q12778
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2308
Test Specificity Detects human FoxO1/FKHR in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human FoxA1 (aa 253-472), B1 (aa 191-324), C1 (aa 208-322), D3 (aa 1-140), F1 (aa 129-354), G1 (aa 283-489), J1 (aa 301-421), J3 (aa 375-552), K1 (aa 493-670), M1 (aa 642-763), O3A (aa 372-673), P1 (aa 231-420), P2 (aa 269-460), P3 (aa 179-279), or P4 (aa 229-410) is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.