Learn More
Description
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-FXN (100μg); Mouse purified polyclonal anti-FXN (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
This nuclear gene encodes a mitochondrial protein which belongs to FRATAXIN family. The protein functions in regulating mitochondrial iron transport and respiration. The expansion of intronic trinucleotide repeat GAA results in Friedreich ataxia. Alternative splicing results in multiple transcript variants.
ELISA
Specifications
Specifications
| Antigen | FXN |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FXN. |
| Gene | FXN |
| Gene Alias | CyaY/FA/FARR/FRDA/MGC57199/X25 |
| Gene Symbols | FXN |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.