Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXYD domain containing ion transport regulator 3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004246 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-246 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-246 Supplier Abnova Corporation Supplier No. H00005349A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant FXYD3.

This gene belongs to a small family of FXYD-domain containing regulators of Na+/K+ ATPases which share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD, and containing 7 invariant and 6 highly conserved amino acids. This gene encodes a cell membrane protein that may regulate the function of ion-pumps and ion-channels. This gene may also play a role in tumor progression. Alternative splicing results in multiple transcript variants encoding distinct isoforms

Sequence: NDLEDKNSPFYYDWHSLQVGGLICAGVLCAMGIIIVMSAKCKCKFGQKSGHHPGETPPLITPGSAQS

Specifications

Antigen FXYD domain containing ion transport regulator 3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant FXYD3.
Formulation 50% glycerol
Gene FXYD3
Gene Accession No. BC005238
Gene Alias MAT-8/MAT8/MGC111076/PLML
Gene Symbols FXYD3
Host Species Mouse
Immunogen FXYD3 (AAH05238, 21 a.a. ∼ 87 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5349
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.