Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD67 Mouse anti-Human, Rat, Mouse, Clone: 1G10.2, MilliporeSigma™
GSA_VA

Catalog No. MAB5406MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5406MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5406MI Supplier MilliporeSigma Supplier No. MAB5406
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This Anti-GAD Antibody, clone 1G10.2 is validated for use in immunohistochemistry, immunohistochemistry (paraffin), western blotting for the detection of GAD67. This Anti GAD antibody has more than 75 product citations.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GAD67
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1G10.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2a in buffer containing 0.1% sodium azide
Gene Accession No. NM_000817.2
Gene Symbols GAD1, SCP, GAD-67, FLJ45882, GAD67, GAD, EC 4.1.1.15
Host Species Mouse
Immunogen Recombinant GAD67 protein
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.