Learn More
Advanced ImmunoChemical, Inc. ANTI-GAPDH MAB
Supplier:  Advanced ImmunoChemical, Inc. 2RGM21 MG
Encompass_Preferred
MONOCLONAL ANTIBODIES/MAbs in vitro:6C5ccMAbs in vivo:4G5Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with rabbit muscle GAPDH.Specificity:Cross-reaction of MAbs in Western blotting with GAPDH from different animal species. MAb isotypes:IgG1 for all MAb 6C5cc, 4G5Applications:GAPDH immunoassay, Western blotting, and immunocytochemistry. Purification:Protein A chromatographyPresentation:PBS, pH 7.4, 0.09% sodium azide (NaN3) Storage Conditions:+4 °C (+2 …. +8 Celsius allowed)
Catalog No. NC3566227
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.