Learn More
ABclonal Technology GEN1 Polyclonal Antibody, 100UL, Validated in:WB
SDP
Supplier:  ABclonal Technology A10001
Encompass_Preferred
Immunogen: Recombinant fusion protein containing a sequence corresponding to amino acids 669-908 of human GEN1 (NP_872431.3).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse•Validated Applications:WB•Swiss Prot: Q17RS7•Gene ID:348654•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-130-9862