Learn More
Green Fluorescent Protein Chicken anti-Vertebrates, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Green Fluorescent Protein |
| Applications | Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified chicken immunoglobulin in TBS, pH 7.5 with 0.02% sodium azide. |
| Host Species | Chicken |
| Immunogen | A recombinant green fluorescent protein (GFP) containing a 6-his tag, was highly purified by affinity chromatography on Nickel column followed by chromatography on a P-100 gel filtration column. The resulting product was present as only a singl |
| Purification Method | Ammonium sulfate precipitation |
| Quantity | 250 μg |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.