Learn More
MBL International Corporation Anti-GFP (Green Fluorescent Protein) mAb-Magnetic Beads (Monoclonal Antibody)
SDP
Supplier:  MBL International Corporation D15311
Encompass_Preferred
Clonality:Monoclonal; Application:IP; Conjugate:Magnetic Beads; Size:20 Tests (Slµrry: 1 mL); Target:GFP; Monoclonal antibody of 20 tests targeting GFP for IPP conjugated to magnetic beads.GFP Monoclonal Antibody.
Catalog No. NC1498241
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.