Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTPase, IMAP family member 4, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004662 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-662 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-662 Supplier Abnova Corporation Supplier No. H00055303A01
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant GIMAP4.

This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. The encoded protein of this gene may be negatively regulated by T-cell acute lymphocytic leukemia 1 (TAL1). In humans, the IAN subfamily genes are located in a cluster at 7q36.1. [provided by RefSeq

Sequence: RYTEEEHKATEKILKMFGERARSFMILIFTRKDDLGDTNLHDYLREAPEDIQDLMDIFGDRYCALNNKATGAEQEAQRAQLLGLIQRVVRENKEGCYTNR

Specifications

Antigen GTPase, IMAP family member 4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant GIMAP4.
Formulation 50% glycerol
Gene GIMAP4
Gene Accession No. NM_018326
Gene Alias FLJ11110/HIMAP4/IAN1/IMAP4/MSTP062/hIAN1
Gene Symbols GIMAP4
Host Species Mouse
Immunogen GIMAP4 (NP_060796, 125 a.a. ∼ 224 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 55303
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.