Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLI-Kruppel family member GLI4, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89005445 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-005-445 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-005-445 Supplier Abnova Corporation Supplier No. H00002738B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human GLI4 protein.

Sequence: MAALGDIQESPSVPSPVSLSSPGTPGTQHHEPQLHLHGHQHGSPGSSPKVLSQPSDLDLQDVEEVEIGRDTFWPDSEPKPEQAPRSPGSQAPDEGAGGALRSLLRSLPRRARCSAGFGPESSAERPAGQPPGAVPCAQPRGAWRVTLVQQAAAGPEGAPERAAELGVNFGRSRQGSARGAKPHRCEACGKSFKYNSLLLKHQRIHTGEKPYACHECGKRFRGWSGFIQHHRIHTGEKPYECGQCGRAFSHSSHFTQHLRIHNGEKPYKCGECGQAFSQSSNLVRHQRLHTGEKPYACSQCGKAFIWSSVLIEHQRIHTGEKPYECSDCGKAFRGRSHFFRHLRTHTGEKPFACGACGKAFGQSSQLIQHQRVHYRE

Specifications

Antigen GLI-Kruppel family member GLI4
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human GLI4 protein.
Formulation No additive
Gene GLI4
Gene Accession No. NM_138465.3
Gene Alias HKR4
Gene Symbols GLI4
Host Species Mouse
Immunogen GLI4 (NP_612474.1, 1 a.a. ∼ 376 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2738
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.