Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR-2 Rabbit anti-Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB10529MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10529MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10529MI Supplier MilliporeSigma Supplier No. AB10529
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-GluR-2 Antibody, C-terminus detects level of GluR-2 and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR-2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum containing 0.05% sodium azide.
Gene Symbols HBGR2; GLUR2; GLURB; GluR-K2; GluR-B; GluR-2; gluR-B; GRIA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to the C-terminus of rat GluR-2.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001077280
Target Species Mouse, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.