Learn More
Glutamate Receptor 2 Mouse anti-Mouse, Rat, Monkey, Canine, Clone: 6C4, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Glutamate Receptor 2 |
| Applications | ELISA, Immunocytochemistry, Immunohistochemistry, Radioimmune Assays (RIA), Western Blot |
| Classification | Monoclonal |
| Clone | 6C4 |
| Conjugate | Unconjugated |
| Formulation | Purified mouse immunoglobin IgG2a liquid in buffer containing PBS, no preservative. |
| Gene Accession No. | NP_058957 |
| Gene Symbols | Gria2, GLUR2, GLURB, GluR-2, GluR-B, GluR-K2 |
| Host Species | Mouse |
| Immunogen | Recombinant fusion protein TrpE-GluR2 (putative N-terminal portion, amino acids 175-430). |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.