Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyceraldehyde-3-Phosphate Dehydrogenase Mouse anti-Canine, Human, Mouse, Rat, Rabbit, Fish, Feline, Pig, Clone: 6C5, MilliporeSigma™
GSA_VA

Catalog No. MAB374MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB374MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB374MI Supplier MilliporeSigma Supplier No. MAB374
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5 is a well published and extensively characterized monoclonal antibody. This purified mAb detects Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) and has been published and validated for use in ELISA, immunoprecipitation, immunocytochemistry, immunofluoresence, immunohistochemistry and western blotting.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Glyceraldehyde-3-Phosphate Dehydrogenase
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6C5
Conjugate Unconjugated
Formulation Purified Mouse monoclonal IgG1 in PBS, pH 7.4 with 0.1% sodium azide.
Gene Accession No. NP_002037
Gene Symbols GAPDH, G3PDH, GAPD, G3PD
Host Species Mouse
Immunogen Glyceraldehyde-3-phosphate dehydrogenase from rabbit muscle.
Purification Method Protein A purfied
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Canine, Feline, Fish, Human, Mouse, Pig, Rabbit, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.