Learn More
Description
Host Species: Rat
Clone BVD2-21C11
Isotype IgG2a, κ
Species Reactivity Hu
Immunogen Recombinant human GM-CSF
Flow Cytometry, Intracellular Staining
Specifications
Specifications
| Antigen | GM-CSF |
| Classification | Monoclonal |
| Clone | BVD2-21C11 |
| Conjugate | PE-CF594 |
| Host Species | Rat |
| Immunogen | Recombinant human GM-CSF |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.