Learn More
ECM Bioscience GM130 (C-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience GM3421
Encompass_Preferred
Clone M342 was generated from a recombinant protein corresponding to amino acid residues in the C-terminal region of human GM130. This sequence has high homology to similar regions in rat and mouse GM130. Accession #Q08379. This antibody detects a 130 kDa* protein corresponding to the apparent molecular mass of GM130 on SDS-PAGE immunoblots of mouse C2C12 cells. In immunocytochemistry, anti-GM130 specifically stains Golgi in mouse C2C12 cells. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = GOLGA2, golgi autoantigen
Catalog No. 50-206-8723
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.