Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM130 Mouse, Unlabeled, Clone: 35/GM130, BD

Catalog No. bdb610822 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610822 50 μg
BDB610823 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610822 Supplier BD Biosciences Supplier No. 610822
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Maturation and post-translational modification of proteins occurs after their biosynthesis at the endoplasmic reticulum and their transport through the Golgi apparatus. The process involves the transport of vesicles carrying the proteins through a vectorial process of vesicle budding and fusion from the cis-compartment to the medial-compartment and the trans-compartment of the Golgi apparatus. The detergent insoluble fraction of the Golgi is named “matrix” and is required for proper morphology of the Golgi membranes. GM130 (Golgi matrix protein of 130kDa) is a protein isolated from the Triton™ X-100-insoluble Golgi matrix and peripherally associated with the cis compartment, as demonstrated by co-localization with syntaxin5. GM130 is homologous to the Golgi autoantigen golgin 95. GM130 interacts through its N-terminal domain with p115 and with the Golgi membranes at the C-terminal portion. Furthermore, the mitotic phosphorylation of GM130 blocks the interaction with p115. Thus, GM130 appears to function as a structural element of the Golgi apparatus that also provides attachment sites for membranes and other Golgi proteins. The 35/GM130 monoclonal antibody recognizes GM130, regardless of phosphorylation status.

Host Species: Mouse
Clone: 35
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Rat GM130 aa. 869-982

Immunofluorescence, Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen GM130
Applications Western Blot
Classification Monoclonal
Clone 35/GM130
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat GM130 aa. 869-982
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.