Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMAP-210 (Trip230) Mouse anti-Human, Unlabeled, Clone: 15, BD

Catalog No. BDB611712 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611712 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611712 Supplier BD Biosciences Supplier No. 611712

Mouse Monoclonal Antibody

Trip230 was identified as a protein that interacts with retinoblastoma protein (Rb) and the thyroid hormone receptor (TR). The structure of Trip230 includes five coiled-coil segments separated by non-helical linkers, N-terminal and C-terminal leucine zipper domains, and multiple phosphorylation sites. Trip230 is ubiquitously expressed and localized to the Golgi. During cell cycle progression, a significant portion of Trip230 translocates to the nucleus. In addition, activation of TR with thyroid hormone (T3) leads to phosphorylation of Trip230, as well as Trip230 translocation from the Golgi to the nucleus. Interestingly, Trip230 has also been identified as a Golgi microtubule-associated protein of 210kDa (GMAP-210). In vitro , GMAP-210 can bind to the minus end of α-tubulin and γ-tubulin via its C-terminal region, while the N-terminal region is involved in Golgi binding. Overexpression of GMAP-210 leads to enlargement of the Golgi apparatus and alterations in the microtubule cytoskeleton. Thus, GMAP-210/Trip230 is thought to function both as a TR coactivator and as a microtubule-binding protein that anchors the Golgi to the microtubule cytoskeleton.

Host Species: Mouse
Clone: 15
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human GMAP-210 aa. 159-365

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen GMAP-210 (Trip230)
Applications Western Blot
Classification Monoclonal
Clone 15
Concentration 250μg/mL
Conjugate Unconjugated
Description Trip230
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human GMAP-210 aa. 159-365
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.