Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDP-mannose pyrophosphorylase A, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018152 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-152 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-152 Supplier Abnova Corporation Supplier No. H00029926D01P
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human GMPPA protein.

This gene is thought to encode a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides. [provided by RefSeq

Sequence: MLKAVILIGGPQKGTRFRPLSFEVPKPLFPVAGVPMIQHHIEACAQVPGMQEILLIGFYQPDEPLTQFLEAAQQEFNLPVRYLQEFAPLGTGGGLYHFRDQILAGSPEAFFVLNADVCSDFPLSAMLEAHRRQRHPFLLLGTTANRTQSLNYGCIVENPQTHEVLHYVEKPSTFISDIINCGIYLFSPEALKPLRDVFQRNQQDGQLEDSPGLWPGAGTIRLEQDVFSALAGQGQIYVHLTDGIWSQIKSAGSALYASRLYLSRYQDTHPERLAKHTPGGPWIRGNVYIHPTAKVAPSAVLGPNVSIGKGVTVGEGVRLRESIVLHGATLQEHTCVLHSIVGWGSTVGRWARVEGTPSDPNPNDPRARMDSESLFKDGKLLPAITILGCRVRIPAEVLILNSIVLPHKELSRSFTNQIIL

Specifications

Antigen GDP-mannose pyrophosphorylase A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human GMPPA protein.
Formulation PBS with no preservative; pH 7.4
Gene GMPPA
Gene Accession No. NM_013335.2
Gene Symbols GMPPA
Host Species Rabbit
Immunogen GMPPA (NP_037467.2, 1 a.a. ∼ 420 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29926
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.