Learn More
R&D Systems Anti-Goat Detection Module, Anti-Goad Secondary HRP Antibody, Luminol-S, Peroxide, Milk-Free Antibody Biluent, and Streptavidin
SDP
Catalog No. NC1409164 Shop All Novus Products
Encompass
Compatible with Jess, Abby, Wes, Peggy Sue and Sally Sue systems. Module includes: Anti-Goat Secondary HRP Antibody, Luminol-S, Peroxide, Milk-free Antibody Diluent (use to dilute goat primary antibody and as blocking agent), and Streptavidin-HRP.
Catalog No. NC1409164 Supplier R&D Systems Supplier No. DM006/DEL
Customers require a valid business account associated to their profile to access pricing and ordering options.
Compatible with Jess, Abby, Wes, Peggy Sue and Sally Sue systems. Module includes: Anti-Goat Secondary HRP Antibody, Luminol-S, Peroxide, Milk-free Antibody Diluent (use to dilute goat primary antibody and as blocking agent), and Streptavidin-HRP.