Learn More
Rockland Immunochemicals TrueBlot Anti-Goat IgG Magnetic Beads 5.0 mL
SDP
Supplier:  Rockland Immunochemicals 00184450
Encompass_Preferred
TrueBlot Magnetic Beads are uniform non-aggregating super-paramagnetic beads consisting of a ferric oxide core functionalized with various silane groups. The super-paramagnetic nanoparticles are coupled with a biomolecule such as rabbit Anti-goat IgG and are specifically designed tested and quality controlled for isolation and purification of goat IgG and immunoprecipitation methods using manual or automatic platforms. This antibody binds the heavy chain of goat IgG and is suitable for immunoassays that utilize a goat IgG primary polyclonal antibody. Cell separation and sorting can be achieved using a goat IgG antibody to defined cell surface antigens. The beads have a large surface area with high capture efficiencies. The beads are in suspension and will settle upon storage. Prior to use mix the vial gently (do not vortex) to ensure delivery of proper bead volume. Bead mean diameter is ~0.5 μm bead concentration is 5 mg/mL.
Catalog No. 50-194-3751
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.