Learn More
Novus Biologicals Anti-Goat Secondary HRP Antibody
SDP
Supplier:  Novus Biologicals 0435222
Encompass
Donkey Anti-Goat Secondary HRP Conjugate (ready-to-use reagent) for use on Jess, Abby, Wes, Peggy Sue and Sally Sue systems. ProteinSimple recommends the use of our Milk-free Antibody Diluent (043-524) to dilute goat primary and as a blocking agent.
Catalog No. NC1475672