Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Golgin-84 Mouse, Unlabeled, Clone: 26, BD

Catalog No. BDB611382 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611382 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611382 Supplier BD Biosciences Supplier No. 611382

Mouse Monoclonal Antibody

The Golgi apparatus is a complex and dynamic organelle that functions in protein sorting and modification. Numerous structural and regulatory proteins are involved in the budding, docking, and fusion of Golgi-directed vesicles. Golgin-84 is an integral membrane protein associated with the Golgi. Sequence analysis of the C-terminal region of Golgin-84 demonstrates a 14 residue region extending into the lumen of the Golgi, a membrane insertion sequence, and a Golgi retention signal. The large N-terminal cytoplasmic region contains a coiled-coil domain that is required for dimerization and two leucine zipper domains. Golgin-84 is ubiquitously expressed, but is abundant in testis. Golgin-84 has sequence homology to coiled-coil containing myosin family members and to several other Golgi proteins, including trans-Golgi p230 and gigantin. Trans-Golgi p230 has been implicated in the biogenesis of specific Golgi vesicles, while gigantin is implicated in the tethering of non-clathrin coated vesicles and in the anchoring of adjacent Golgi cisternae. Thus, Golgin-84 may have similar roles in vesicle biogenesis, docking, and fusion at the Golgi interface.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Golgin-84
Applications Western Blot
Classification Monoclonal
Clone 26
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Golgin-84 aa. 510-713
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.