Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gp91[phox] Mouse, Unlabeled, Clone: 53, BD

Catalog No. BDB611414 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611414 50 μg
BDB611415 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611414 Supplier BD Biosciences Supplier No. 611414
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Although dormant in resting granulocytes, macrophages, and B lymphocytes, the neutrophil respiratory burst oxidase (NADPH-oxidase) generates superoxide and secondary oxygen-derived toxic products in response to bacteria or a variety of soluble stimuli. It is an integral membrane cytochrome, b558, which consists of two subunits, gp91[phox] and p21[phox]. Upon stimulation, cytochrome b558 forms a complex with cytosolic proteins, p67[phox], p47[phox], p40[phox], and rac2 and produces superoxide anions in a NADPH-dependent
manner. In chronic granulomatous disease (CGD), severe recurrent bacterial and fungal infections result from defective NADPH-oxidase activity. The majority of CGD cases are caused by a defective gp91[phox] gene. gp91[phox], implicated as a docking site for p47[phox], is membrane glycoprotein with multiple N-terminal hydrophobic domains and a hydrophilic C-terminus. The expression of gp91[phox] is restricted to terminally differentiated phagocytes and B lymphocytes. This cell type- and developmental stage-specific expression may be
controlled by transcriptional activators, such as PU.1 and YY1, and transcriptional repressors, such as CCAAT displacement protein (CDP). Endogenous mouse gp91phox shows 58kDa band, instead of 91kDa observed in human. Both mouse and human deglycosylated gp91 are 54kDa. This difference may be due to less glycosylation sites in the mouse sequence.

Host Species: Mouse
Clone: 53
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Mouse gp91[phox] aa. 450-556
Formula Weight [Chemical]: 58kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen gp91[phox]
Applications Western Blot
Classification Monoclonal
Clone 53
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse gp91[phox] aa. 450-556
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.