Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gp91-phox Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 07024MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-024-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-024-MI Supplier MilliporeSigma Supplier No. 07024
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-gp91-phox Antibody is an antibody against gp91-phox for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gp91-phox
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol. Liquid at −20°C.
Gene Symbols CGD; CGD91-phox; EC 1.-.-.-; GP91-1; GP91-PHOX; GP91PHOX; NOX2; OTTHUMP00000031915; gp91-1; gp91-phox; p91-PHOX
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 548-560 of human gp91-phox (KQSISNSESGPRG). The immunizing sequence is identical in mouse, and it shares 11/13 sequence homology in bovine and 10/13 in porcine.
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000397
Target Species Human
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.