Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granzyme B Mouse anti-Human, PE-CF594, Clone: GB11, BD

Catalog No. BDB562462 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562462 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562462 Supplier BD Biosciences Supplier No. 562462
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. Antibody was conjugated with Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed. Granzyme B is stored in the granules of cytotoxic T lymphocytes and NK cells along with the pore-forming protein perforin. Granzyme B acts on specific substrates including caspase-3, -7, -9, and -10 which in turn give rise to enzymes that mediate apoptosis.

Host Species: Mouse
Clone: GB11
Isotype: IgG1, κ
Species Reactivity: Human
Immunogen: Human Granzyme B

  • Involved in the hydrolysis of extracellular matrix components
  • PE-CF594 can be excited by the blue (488nm), green (532nm) and yellow-green (561nm) lasers and can be detected with the same filter set as PE-Texas Red

Intracellular Staining, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Granzyme B
Applications Flow Cytometry
Classification Monoclonal
Clone GB11
Concentration 5μL
Conjugate PE-CF594
Description GZMB; Granzyme-2; CCPI; CGL1; CSPB; CTLA1; CTSGL1; GRB; HLP; SECT
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Granzyme B
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.