Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granzyme K Mouse anti-Human, Clone: GM6C3, Invitrogen™

Catalog No. 01670168 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-168 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-168 Supplier Thermo Scientific Supplier No. MA117755

Item Discontinued This product is either new or inactive, and cannot be ordered online.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This antibody recognizes native Granzyme K protein expressed in transfected mammalian cells. This antibody can be used in a Sandwich ELISA as a detection antibody with Product No. MA1-17754 as a capture antibody.

Granzyme K (GrK) belongs to a family of trypsin-like serine proteases localised in the cytotoxic granules of activated T cells and NK cells. It encodes a 28 kDa serine protease whose gene is located on chromosome 5q11-12 close to the granzyme A-encoding gene. Like granzyme A, it has a trypsin-like specifity cleaving at the basic residues arginine and lysine. Granzyme K triggers rapid cell death independently of caspase activation with single-stranded DNA nicks and is primarily expressed in thymus, lung, spleen and peripheral blood leukocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Granzyme K
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone GM6C3
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.2
Gene GZMK
Gene Accession No. P49863
Gene Alias GrK
Gene Symbols GZMK
Host Species Mouse
Immunogen cDNA encoding human Granzyme K
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3003
Target Species Human
Content And Storage Store at 4°C short term. For long term storage store at -20°C. Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.