Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRK 2/3 (ΒARK 1/2) Mouse anti-Human, Rat, Mouse, Bovine, Chicken, Pig, Rabbit, Clone: C5/1.1, MilliporeSigma™

Catalog No. 05465MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-465-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-465-MI Supplier MilliporeSigma Supplier No. 05465
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect GRK 2/3 (βARK 1/2) using this Anti-GRK 2/3 (βARK 1/2) Antibody, clone C5/1.1 validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRK 2/3 (βARK 1/2)
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone C5/1.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl containing 0.05% sodium azide. Frozen solution.
Gene Accession No. NM_001619, NM_005160
Gene Symbols ADRBK1, Beta-ARK-1, GRK2, BARK, FLJ16718, BETA-ARK1, BARK1
Host Species Mouse
Immunogen Partial GST fusion protein corresponding to the GRK3 (βARK2) carboxyl terminus (a.a. 467-688). This immunogen shares homology with GRK2 (βARK1). The minimal epitope recognized by the antibody is a.a. 483-485. Clone C5/1.1.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Chicken, Human, Mouse, Pig, Rabbit, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.