Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GS27 Mouse, Unlabeled, Clone: 25, BD

Catalog No. BDB611034 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611034 50 μg
BDB611035 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611034 Supplier BD Biosciences Supplier No. 611034

Mouse Monoclonal Antibody

Eukaryotic protein trafficking involves the packaging of target molecules into membranous vesicles that bud from a donor compartment, travel to a specific destination, fuse, and release their components into an acceptor compartment. Components of both the vesicle and the synaptic plasma membrane interact to form a fusion complex that mediates vesicle docking and fusion. This fusion complex contains NSF (N-ethyl-maleimide-sensitive factor), SNAPs (soluble NSF attachment proteins), and receptor proteins (SNAREs) that include synaptobrevin, synaptotagmin, syntaxin, and SNAP-25 (synaptosome-associated protein of 25kDa). Interactions between vesicle SNAREs and target membrane SNARES mediates the specificity of docking. Along one pathway of protein trafficking, the Golgi apparatus receives proteins from the ER. These proteins move from cis-Golgi to trans-Golgi, through a stack of cisternae, towards the trans-Golgi network. From here, they are delivered to their proper destination in the cell. GS27 (Golgi SNARE of 27kDa) (membrin) is a Golgi-associated SNARE. It functions in medial-to-trans-Golgi protein movement.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen GS27
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 25
Concentration 250μg/mL
Conjugate Unconjugated
Description Golgi SNARE of 27 kDa; Membrin
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human GS27 aa. 5-124
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.