Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GS28 Mouse, Unlabeled, Clone: 1, BD

Catalog No. BDB611184 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611184 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611184 Supplier BD Biosciences Supplier No. 611184
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Most vesicular fusion events require the function of N-ethylmaleimide-sensitive factor (NSF) and the soluble NSF attachment proteins (SNAPs). Their function is dependent upon the SNAP receptor (SNARE) family of proteins. The specificity of vesicle docking and fusion is mediated by specific interactions between v-SNAREs on vesicles and t-SNAREs on target membranes. GS28 (Golgi SNARE with a size of 28kDa) is a SNARE that associates with the cis-Golgi and participates in trafficking between the ER and the Golgi and between Golgi
compartments. The majority of GS28, the first 230 and 250 aa, is thought to be anchored to the membrane via the C-terminal hydrophobic tail, which is formed by the remaining 20 aa. GS28 and syntaxin 5, another SNARE, exist as a protein complex in the Golgi and this complex is important for the function of both proteins in ER-Golgi transport. The GS28/syntaxin 5 complex can be dissociated by α-SNAP and NSF. In addition, GS28 is thought to interact with α-SNAP when the GS28/syntaxin 5 complex is dissociated. Thus, GS28 is a SNARE protein that mediates, in complex with syntaxin 5, transport within the Golgi and between Golgi and ER.

Host Species: Mouse
Clone: 1
Isotype: IgG2a
Species Reactivity: Mouse
Immunogen: Rat GS28 aa. 3-108
Formula Weight [Chemical]: 28kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen GS28
Applications Western Blot
Classification Monoclonal
Clone 1
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat GS28 aa. 3-108
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.