Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC, Human, IP-WB Antibody Pair (PW4), Abnova™

Catalog No. 89302062 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-302-062 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-302-062 Supplier Abnova Corporation Supplier No. H00145258PW4
Add to Cart
Add to Cart

Includes

Antibody pair for IP: rabbit polyclonal anti-GSC (300μL), Antibody pair for WB: mouse purified polyclonal anti-GSC (50μg)

For immunoprecipitation and to detect the precipitated protein in western blot.

Specifications

Antigen GSC
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene GSC
Gene Symbols GSC
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145258
Includes Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-GSC (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-GSC (50 ug)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.