Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GTF3C5, Human, IP-WB Antibody Pair, Abnova™

Catalog No. 89301350 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-301-350 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-301-350 Supplier Abnova Corporation Supplier No. H00009328PW1
Add to Cart
Add to Cart

Includes

Antibody pair for IP: mouse monoclonal anti-GTF3C5 (300μg), Antibody pair for WB: mouse monoclonal anti-GTF3C5 (50μg)

For immunoprecipitation and to detect the precipitated protein in western blot.

Specifications

Antigen GTF3C5
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene GTF3C5
Gene Alias FLJ20857/TFIIIC63/TFIIICepsilon/TFiiiC2-63
Gene Symbols GTF3C5
Host Species Mouse
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9328
Includes Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-GTF3C5 (300 ug) 2. Antibody pair for WB: mouse monoclonal anti-GTF3C5 (50 ug)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.