Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Guinea Pig IgG, Isotype Control, Novus Biologicals™
SDP

Catalog No. NBP1970365 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1970365 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP1970365 Supplier Novus Biologicals Supplier No. NBP1970365MG
Only null left
Add to Cart
Add to Cart

Guinea Pig Antibody has been used in 3 publications

IgG specifically detects IgG in Guinea Pig samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, SDS-Page.

Specifications

Antigen IgG
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), SDS-Page
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, SDS-Page 1:100 - 1:2000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: None. with 0.01% Sodium Azide
Gene Alias immunoglobulin heavy constant gamma 1 (G1m marker)
Host Species Guinea Pig
Purification Method Ion exchange chromatography
Quantity 5 mg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Reconstitution Reconstitute with 0.5 ml deionized water (or equivalent) to desired concentration.
Target Species Guinea Pig
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.