Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

histidyl-tRNA synthetase 2, mitochondrial (putative), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004561 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-561 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-561 Supplier Abnova Corporation Supplier No. H00023438A01
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant HARSL.

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is an enzyme belonging to the class II family of aminoacyl-tRNA synthetases. Functioning in the synthesis of histidyl-transfer RNA, the enzyme plays an accessory role in the regulation of protein biosynthesis. The gene is located in a head-to-head orientation with HARS on chromosome five, where the homologous genes share a bidirectional promoter. [provided by RefSeq

Sequence: PLLGLLPRRAWASLLSQLLRPPCASCTGAVRCQSQVAEAVLTSQLKAHQEKPNFIIKTPKGTRDLSPQHMVVREKILDLVISCFKRHGAKGMDTPAFELKETLTEKYGE

Specifications

Antigen histidyl-tRNA synthetase 2, mitochondrial (putative)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant HARSL.
Formulation 50% glycerol
Gene HARS2
Gene Accession No. NM_012208
Gene Alias HARSL/HARSR/HO3
Gene Symbols HARS2
Host Species Mouse
Immunogen HARSL (NP_036340, 2 a.a. ∼ 110 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 23438
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.