Learn More
ACROBiosystems Anti-HBcAg Antibody IgG Pair
Supplier:  ACROBiosystems RASP165100UG
Encompass_Preferred
Hepatitis B core antigen (HBcAg) is a single polypeptide with a molecular weight of 18848u. HBcAg plays an important role in HBV infection. It can reflect the presence of Dane particles in serum and the replication of HBV in liver and can cooperate and complement with other HBV serological markers. Only because its antibody has a strong affinity and can rapidly combine with HBcAg in serum to form immune complexes it is difficult to measure free HBcAg in serum. Double antibody sandwich ELISA is commonly used to detect HBcAg.
Catalog No. 50-253-4260
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.