Learn More
Invivogen Anti-hCD19-CD3 Bispecific Antibody, Human CD19 and CD3 Targeting, Azide-Free, 50 µg
SDP
Supplier:  Invivogen BIMABCD19CD305
Encompass
The Anti-hCD19-CD3 bispecific antibody from InvivoGen is an innovative tool for immunotherapy research, designed to engage T cells for targeted attack on CD19-positive cells. With unique attributes like its azide-free, lyophilized formulation and high purity (>90%), this monoclonal antibody enables effective studies of cell activation and proliferation. Its specificity for human CD19 and CD3, coupled with no cross-reactivity with other species, distinguishes it from competitors.
Catalog No. NC2456657
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.