Learn More
Immunology Consultants Monoclonal Mouse anti-hCG beta Antibody Clone#110, 1.0 mg
SDP
Supplier:  Immunology Consultants MGCB80A110
Encompass_Preferred
Monoclonal antibody specific to the beta chain of human chorionic gonadotropin.  There is no cross-reactivity with LH, TSH, or FSH, making them suitable for diagnosing early pregnancy, with a detection limit of below 10 mIU/ml.
Catalog No. NC2734267
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.