Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B Virus Mouse anti-Human, Clone: 10E11, MilliporeSigma™

Catalog No. MAB16989MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB16989MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB16989MI Supplier MilliporeSigma Supplier No. MAB16989
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Hepatitis B Virus Antibody, core antigen (ayw), clone 10E11, a.a. 2-10 detects level of Hepatitis B Virus and has been published and validated for use in ELISA, immunofluoresence, immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis B Virus
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 10E11
Conjugate Unconjugated
Formulation Ascitic fluid. Liquid in DMEM, containing 0.1% sodium azide.
Host Species Mouse
Immunogen Recombinant purified HBcAg from E.coli, SDS-denatured
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.