Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HER-2/neu Mouse anti-Human, PE, Clone: NEU 24.7, BD

Catalog No. BDB340552 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340552 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340552 Supplier BD Biosciences Supplier No. 340552
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-HER-2/neu recognizes a single protein of 185kDa from HER-2 transfected cells.The HER-2/neu antigen, product of the c-erbB2 gene, is a surface marker on some breast cancer cells. Amplification and/or overexpression of HER-2/neu occurs in multiple human malignancies. Overexpression is reported to be strongly correlated with progression of human ovarian and breast carcinomas. Anti-HER-2/neu stains the external domain of this protein on cells such as the strongly expressing line SKBR3 and the weakly expressing line MCF7.

Applications:

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen HER-2/neu
Applications Flow Cytometry
Classification Monoclonal
Clone NEU 24.7
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen SKBR3 human breast carcinoma cells
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.