Learn More
Description
Comprend
Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-HIP1 (100μg); Detection antibody: mouse monoclonal anti-HIP1, IgG2a Kappa (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Host Species: Rabbit, Mouse
Isotype: IgG, IgG2a, κ
Cross Reactivity: Human
ELISA Pair
Spécifications
Spécifications
| Antigen | HIP1 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human HIP1. |
| Gene | HIP1 |
| Gene Alias | ILWEQ/MGC126506 |
| Gene Symbols | HIP1 |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Afficher plus de résultats |
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.