Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2B Rabbit anti-Chicken, Human, Polyclonal, MilliporeSigma™

Catalog No. 07371MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-371-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-371-MI Supplier MilliporeSigma Supplier No. 07371
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Histone H2B Antibody, Cat. No. 07-371, is a highly specific rabbit polyclonal antibody that targets Histone H2B and has been tested in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H2B
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.02M phosphate buffer, pH 7.6, 0.25M NaCl, and 0.1% sodium azide, and 30% glycerol.
Gene Symbols GL105; H2B; H2B-GL105; H2B.1; H2B.q; H2B/q; H2BFQ; MGC119802; MGC119804; MGC129733; MGC129734; OTTHUMP00000013920
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 118-126 (CG-AVTKYTSSK) of human Histone H2B, with an N-terminal CG added for conjugation purposes.
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_778225
Target Species Chicken, Human, S. cerevisiae, Xenopus species, Yeast
Content And Storage −20°C for up to 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.