Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Anti-Histone H3 (Citrulline R2+R8+R17) Antibody [RM1001], Rabbit, Multiclonal, 1 mg

Catalog No. 502617525
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-261-7525 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-261-7525 Supplier Abcam Supplier No. AB2838631MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Rabbit recombinant multiclonal antibody that is carrier-free and conjugation ready.

This is a rabbit recombinant multiclonal antibody [RM1001] targeting Histone H3 (citrulline R2 + R8 + R17). The product is supplied as a carrier-free liquid.

  • Host Species: Rabbit
  • Clonality: Multiclonal
  • Isotype: IgG
  • Species Reactivity: Human, mouse, and rat
  • Applications: ICC/IF, IP, Dot Blot, Western Blot, Flow Cytometry (Intracellular), and Peptide Array
  • Purification: Affinity purification Protein A
  • Storage Buffer: PBS (pH 7.2 to 7.4)
  • Features: BSA-free and conjugation-ready

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Label or Dye Unconjugated
Target Molecule Histone H3 (citrulline R2, R8, R17) Human, Mouse, Rat
Detection Method Chemiluminescence, Fluorescence
Form Liquid
Detection Location In-Blot Detection
Buffer PBS, pH7.2 to 7.4
Cross Reactivity Human, Mouse, Rat
Formulation PBS, pH 7.2 to 7.4
Quantity 1 mg
For Use With (Application) Western Blot, Flow Cytometry (Intra), Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Dot Blot, Peptide Array
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.