Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-HNF-1-beta Clone: 12A5.1,

Catalog No. MABE971MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE971MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE971MI Supplier MilliporeSigma™ Supplier No. MABE971
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects HNF-1-beta Clone: 12A5.1 in Human, Rat samples, and it is validated for Immunohistochemistry, Western Blotting

Hepatocyte nuclear factor 1-beta (UniProt P35680; also known as HNF-1-beta, HNF-1B, Homeoprotein LFB3, TCF-2, Transcription factor 2, Variant hepatic nuclear factor 1, vHNF1) is encoded by the HNF1B (also known as TCF2, VHNF1) gene (Gene ID 6928) in human. HNF-1-beta is involved in transcriptional and functional regulation of the liver and biliary system, as well as of the kidneys, urogenital tract and pancreatic β-cells. HNF-1-beta deficiency due to HNF1B mutations is a well known cause of maturity-onset diabetes of the young 5 (MODY5), also referred to as RCAD (renal cysts and diabetes) due to the frequent co-occurrence of renal cysts and/or other inborn urogenital abnormalities. HNF-1-beta is expressed in the gall bladder, liver and intrahepatic bile ducts of mouse embryos and in liver of adult mice. HNF-1-beta is shown to co-regulate morphogenesis of the biliary system and HNF1B knockout mice suffer from severe neonatal cholestasis and jaundice due to the abnormally developed gall bladder and dysfunctional intrahepatic bile ducts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HNF-1-beta
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 12A5.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P35680
Gene Symbols HNF1B, TCF2
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the Internal (N-terminal half) of human HNF-1-beta.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_000449
Test Specificity Expected to react with isoform 1 (A) and isoform 3 (C). Reactivity toward isoform 2 (B) and isoform 4 has not been determined.
Target Species Human, Rat
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.