Learn More
Abcam Anti-HNF1 alpha Antibody, Rabbit Polyclonal, 50 mg
Description
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Product Type | CHAPS |
| Format | Vial |
| Label or Dye | Unconjugated |
| Quantity | 50 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.