Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXD4 Mouse anti-Rat, Mouse, Clone: 8E11.1, MilliporeSigma™

Catalog No. MABE157MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE157MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE157MI Supplier MilliporeSigma Supplier No. MABE157

Mouse Monoclonal Antibody

Anti-HOXD4 Antibody, clone 8E11.1 is a Mouse Monoclonal Antibody for detection of HOXD4 also known as Homeobox protein Hox-D4 and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOXD4
Applications Western Blot
Classification Monoclonal
Clone 8E11.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_055436
Gene Symbols HOXD4, HOX4B
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human HOXD4.
Purification Method Purified by ion exchange chromatography
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.