Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPV-16 L1 Mouse anti-Human, Unlabeled, Clone: CAMVIR-1, BD

Catalog No. BDB550840 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550840 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550840 Supplier BD Biosciences Supplier No. 550840
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

More than 60 different types of human papilloma viruses (HPVs) have been isolated. The human papilloma viruses encode three late proteins, which are produced only in terminally differentiating keratinocytes, two of which (the L1 and L2 proteins) are struc

Host Species: Mouse
Clone: CamVir-1
Isotype: IgG2a κ
Species Reactivity: Human

Immunohistochemistry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen HPV-16 L1
Applications Immunohistochemistry
Classification Monoclonal
Clone CAMVIR-1
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.