Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp60 Mouse, Unlabeled, Clone: 24/HSP60, BD

Catalog No. p-3970838 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611562 50 μg
BDB611563 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611562 Supplier BD Biosciences Supplier No. 611562
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Heat shock proteins (Hsp) are a set of highly conserved proteins that include constitutively expressed (Hsp60, Hsp70, and Hsp90) and stress-induced (Hsp27 and Hsp72) proteins. Hsp60 is localized to the mitochondria, where it promotes mitochondrial protein folding and facilitates proteolytic degradation of misfolded or denatured proteins. It binds Hsp10, which regulates the substrate binding and ATPase activity of Hsp60. In HeLa and Jurkat mitochondria, Hsp60 associates with caspase-3 to form a complex that dissociates and releases from the mitochondria during apoptosis. In addition, Hsp60 accelerates the maturation of procaspase-3 through its ATP-dependent “foldase” activity. In addition to its role in protein folding, Hsp60 has also been implicated in immune function. In macrophages, its binding to the toll-like receptor-4 complex induces production of TNFα and nitric oxide and stimulation of a proinflammatory response. Thus, the protein folding function of Hsp60 is involved in mitochondrial protein folding in both normal and apoptotic cells, while release of Hsp60 during necrosis is thought to stimulate a proinflammatory response.

Host Species: Mouse
Clone: 24
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Hsp60 aa. 1-200
Formula Weight [Chemical]: 60kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Hsp60
Applications Western Blot
Classification Monoclonal
Clone 24/HSP60
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Hsp60 aa. 1-200
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.